siRNA Details
| Gene Symbol: Agfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107131.1 | NP_001100601.1 | 4 | 722 | ||
| XM_006249140.2 | XP_006249202.1 | 5 | 902 | |||
| XM_006249141.2 | XP_006249203.1 | 5 | 901 | |||
| XM_006249142.2 | XP_006249204.1 | 5 | 903 | |||

Agfg2
Genomic Map