siRNA Details
| Gene Symbol: Cldn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012022.1 | NP_001012022.1 | 1 | 805 | ||
| GenBank mRNA | BC083582.1 | - | 1 | 805 | 7130270 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn4| Gene Symbol: Cldn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012022.1 | NP_001012022.1 | 1 | 805 | ||
| GenBank mRNA | BC083582.1 | - | 1 | 805 | 7130270 | |