siRNA Details
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100679.1 | NP_001094149.1 | 23 | 3985 | ||
GenBank mRNA | BC082040.1 | - | 5 | 1015 | ||
FQ224397.1 | - | 2 | 129 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100679.1 | NP_001094149.1 | 23 | 3985 | ||
GenBank mRNA | BC082040.1 | - | 5 | 1015 | ||
FQ224397.1 | - | 2 | 129 |