siRNA Details
| Gene Symbol: Orai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013982.1 | NP_001014004.1 | 2 | 673 | ||
| GenBank mRNA | BC088225.1 | - | 2 | 673 | 7322176 | |
| EU644749.1 | - | 2 | 588 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orai1| Gene Symbol: Orai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013982.1 | NP_001014004.1 | 2 | 673 | ||
| GenBank mRNA | BC088225.1 | - | 2 | 673 | 7322176 | |
| EU644749.1 | - | 2 | 588 | |||