siRNA Details
| Gene Symbol: Mlec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013983.1 | NP_001014005.1 | 2 | 507 | ||
| GenBank mRNA | BC089839.1 | - | 2 | 507 | 7365814 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlec| Gene Symbol: Mlec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013983.1 | NP_001014005.1 | 2 | 507 | ||
| GenBank mRNA | BC089839.1 | - | 2 | 507 | 7365814 | |