siRNA Details
Gene Symbol: Oasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009681.1 | NP_001009681.1 | 1, 2 | 204 | ||
GenBank mRNA | AY227756.1 | - | 1, 2 | 204 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
