siRNA Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249589.2 | XP_006249651.1 | 18 | 2588 | ||
| XM_006249591.2 | XP_006249653.1 | 18 | 2522 | |||
| XM_006249592.2 | XP_006249654.1 | 17 | 2517 | |||
| XM_008769388.1 | XP_008767610.1 | 18 | 2586 | |||
| XM_008769389.1 | XP_008767611.1 | 17 | 2558 | |||
| XM_222254.8 | XP_222254.5 | 19 | 2658 | |||

Fbrsl1
Genomic Map