siRNA Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107153.1 | NP_001100623.1 | 2 | 264 | ||
| XM_008769337.1 | XP_008767559.1 | 2 | 272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp30| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107153.1 | NP_001100623.1 | 2 | 264 | ||
| XM_008769337.1 | XP_008767559.1 | 2 | 272 | |||