siRNA Details
| Gene Symbol: Tmem119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107155.1 | NP_001100625.1 | 2 | 435 | ||
| XM_006249500.2 | XP_006249562.1 | 2 | 562 | |||
| GenBank mRNA | BC166820.1 | - | 2 | 388 | 8367783 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem119| Gene Symbol: Tmem119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107155.1 | NP_001100625.1 | 2 | 435 | ||
| XM_006249500.2 | XP_006249562.1 | 2 | 562 | |||
| GenBank mRNA | BC166820.1 | - | 2 | 388 | 8367783 | |