siRNA Details
| Gene Symbol: Sart3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107156.1 | NP_001100626.1 | 4 | 714 | ||
| XM_006249501.2 | XP_006249563.1 | 4 | 1033 | |||
| XM_006249502.2 | XP_006249564.1 | 5 | 665 | |||
| XM_008769338.1 | XP_008767560.1 | 4 | 1473 | |||

Sart3
Genomic Map