siRNA Details
| Gene Symbol: Orai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170403.1 | NP_001163874.1 | 3 | 819 | ||
| GenBank mRNA | BC158835.1 | - | 3 | 723 | 7384003 | |
| EU644750.1 | - | 3 | 819 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orai2| Gene Symbol: Orai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170403.1 | NP_001163874.1 | 3 | 819 | ||
| GenBank mRNA | BC158835.1 | - | 3 | 723 | 7384003 | |
| EU644750.1 | - | 3 | 819 | |||