siRNA Details
| Gene Symbol: RGD1310262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107161.1 | NP_001100631.1 | 2 | 240 | ||
| XM_006255249.2 | XP_006255311.1 | 1 | 1551 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1310262| Gene Symbol: RGD1310262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107161.1 | NP_001100631.1 | 2 | 240 | ||
| XM_006255249.2 | XP_006255311.1 | 1 | 1551 | |||