siRNA Details
| Gene Symbol: Optc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107176.1 | NP_001100646.1 | 5 | 1012 | ||
| XM_006249871.2 | XP_006249933.1 | 6 | 1859 | |||
| XM_006249872.2 | XP_006249934.1 | 6 | 2418 | |||
| XM_008769554.1 | XP_008767776.1 | 5 | 2245 | |||

Optc
Genomic Map