siRNA Details
| Gene Symbol: Cyb5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013126.1 | NP_001013144.1 | 8 | 742 | ||
| GenBank mRNA | BC089945.1 | - | 8 | 742 | 7311492 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyb5r1| Gene Symbol: Cyb5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013126.1 | NP_001013144.1 | 8 | 742 | ||
| GenBank mRNA | BC089945.1 | - | 8 | 742 | 7311492 | |