siRNA Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012027.1 | NP_001012027.1 | 7 | 1372 | ||
| XM_008769647.1 | XP_008767869.1 | 7 | 1317 | |||
| GenBank mRNA | BC088467.1 | - | 7 | 1372 | 7127992 | |
| FQ209388.1 | - | 7 | 1392 | |||
| FQ209496.1 | - | 7 | 1333 | |||
| FQ209598.1 | - | 7 | 1358 | |||
| FQ211013.1 | - | 7 | 1396 | |||
| FQ218285.1 | - | 7 | 1386 | |||
| FQ218922.1 | - | 7 | 1402 | |||
| FQ218964.1 | - | 7 | 1353 | |||
| FQ219160.1 | - | 7 | 1391 | |||
| FQ219233.1 | - | 7 | 1357 | |||

Serpinc1
Genomic Map