siRNA Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012027.1 | NP_001012027.1 | 2 | 425 | ||
| XM_008769647.1 | XP_008767869.1 | 2 | 370 | |||
| GenBank mRNA | BC088467.1 | - | 2 | 425 | 7127992 | |
| FQ209388.1 | - | 2 | 443 | |||
| FQ209496.1 | - | 2 | 386 | |||
| FQ209598.1 | - | 2 | 411 | |||
| FQ210275.1 | - | 2 | 408 | |||
| FQ210716.1 | - | 2 | 444 | |||
| FQ210762.1 | - | 2 | 443 | |||
| FQ211013.1 | - | 2 | 449 | |||
| FQ211135.1 | - | 2 | 443 | |||
| FQ218285.1 | - | 2 | 437 | |||
| FQ218922.1 | - | 2 | 454 | |||
| FQ218964.1 | - | 2 | 408 | |||
| FQ219233.1 | - | 2 | 408 | |||

Serpinc1
Genomic Map