siRNA Details
Gene Symbol: Igsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014787.1 | NP_001014787.1 | 3 | 255 | ||
XM_006250286.2 | XP_006250348.1 | 3 | 414 | |||
XM_006250288.2 | XP_006250350.1 | 2 | 356 | |||
XM_006250289.2 | XP_006250351.1 | 2 | 193 | |||
GenBank mRNA | BC092192.1 | - | 3 | 255 | 7312924 |
