siRNA Details
| Gene Symbol: Aim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250335.1 | XP_006250397.1 | 5, 6 | 1236 | ||
| XM_006250336.2 | XP_006250398.1 | 6, 7 | 1680 | |||
| XM_006250339.1 | XP_006250401.1 | 4, 5 | 1051 | |||
| XM_008769802.1 | XP_008768024.1 | 6, 7 | 2895 | |||
| XM_008769803.1 | XP_008768025.1 | 6, 7 | 3845 | |||
| XM_008769804.1 | XP_008768026.1 | 7, 8 | 3223 | |||
| XM_008769805.1 | XP_008768027.1 | 6, 7 | 2832 | |||
| XM_222949.8 | XP_222949.5 | 5, 6 | 3660 | |||

Aim2
Genomic Map