siRNA Details
| Gene Symbol: Mnda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012029.1 | NP_001012029.1 | 7 | 1364 | ||
| GenBank mRNA | BC085891.1 | - | 7 | 1364 | 7192620 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mnda| Gene Symbol: Mnda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012029.1 | NP_001012029.1 | 7 | 1364 | ||
| GenBank mRNA | BC085891.1 | - | 7 | 1364 | 7192620 | |