siRNA Details
| Gene Symbol: Lamb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100841.1 | NP_001094311.1 | 5, 6 | 563 | ||
| XM_008769856.1 | XP_008768078.1 | 5, 6 | 474 | |||
| GenBank mRNA | Y08883.1 | - | 2, 3 | 195 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamb3| Gene Symbol: Lamb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100841.1 | NP_001094311.1 | 5, 6 | 563 | ||
| XM_008769856.1 | XP_008768078.1 | 5, 6 | 474 | |||
| GenBank mRNA | Y08883.1 | - | 2, 3 | 195 | ||