siRNA Details
| Gene Symbol: Cd34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107202.2 | NP_001100672.1 | 3 | 359 | ||
| XM_008769857.1 | XP_008768079.1 | 3 | 649 | |||
| GenBank mRNA | EU448292.1 | - | 3 | 359 | ||
| EU448293.1 | - | 3 | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd34| Gene Symbol: Cd34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107202.2 | NP_001100672.1 | 3 | 359 | ||
| XM_008769857.1 | XP_008768079.1 | 3 | 649 | |||
| GenBank mRNA | EU448292.1 | - | 3 | 359 | ||
| EU448293.1 | - | 3 | 359 | |||