siRNA Details
| Gene Symbol: Cd34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107202.2 | NP_001100672.1 | 7 | 874 | ||
| XM_008769857.1 | XP_008768079.1 | 7 | 1164 | |||
| GenBank mRNA | EU448292.1 | - | 7 | 874 | ||
| EU448293.1 | - | 7 | 874 | |||

Cd34
Genomic Map