siRNA Details
| Gene Symbol: Sept11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107208.2 | NP_001100678.1 | 621 | |||
| GenBank mRNA | BC167769.1 | - | 752 | 7456582 | ||
| EU711414.1 | - | 621 | ||||
| EU711415.1 | - | 621 | ||||
| EU711416.1 | - | 621 | ||||
| EU711417.1 | - | 621 | ||||

Sept11
Genomic Map