siRNA Details
| Gene Symbol: Art3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012034.1 | NP_001012034.1 | 3 | 185 | ||
| XM_006250727.1 | XP_006250789.1 | 3 | 264 | |||
| XM_006250728.1 | XP_006250790.1 | 3 | 264 | |||
| XM_008770026.1 | XP_008768248.1 | 3 | 208 | |||
| XM_008770027.1 | XP_008768249.1 | 3 | 240 | |||
| GenBank mRNA | BC079018.1 | - | 3 | 185 | 7113746 | |
| FQ214932.1 | - | 3 | 125 | |||
| FQ216328.1 | - | 3 | 209 | |||
| FQ223913.1 | - | 3 | 209 | |||
| FQ224878.1 | - | 3 | 196 | |||

Art3
Genomic Map