siRNA Details
| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182952.2 | NP_891997.1 | 2 | 236 | ||
| GenBank mRNA | AF537209.1 | - | 2 | 236 | ||
| AY340181.1 | - | 2 | 215 | |||
| BC168725.1 | - | 2 | 244 | 6919932 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl11| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182952.2 | NP_891997.1 | 2 | 236 | ||
| GenBank mRNA | AF537209.1 | - | 2 | 236 | ||
| AY340181.1 | - | 2 | 215 | |||
| BC168725.1 | - | 2 | 244 | 6919932 | ||