siRNA Details
| Gene Symbol: Txk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024255.1 | NP_001019426.1 | 4 | 460 | ||
| XM_008770141.1 | XP_008768363.1 | 3 | 779 | |||
| XM_008770142.1 | XP_008768364.1 | 3 | 722 | |||
| GenBank mRNA | BC095847.1 | - | 4 | 460 | 7388955 | |

Txk
Genomic Map