siRNA Details
| Gene Symbol: Rbm47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005882.1 | NP_001005882.1 | 2 | 778 | ||
| XM_006250949.2 | XP_006251011.1 | 3 | 1861 | |||
| XM_006250950.2 | XP_006251012.1 | 3 | 2759 | |||
| XM_006250951.2 | XP_006251013.1 | 5 | 1107 | |||
| XM_006250952.2 | XP_006251014.1 | 4 | 1333 | |||
| XM_006250954.2 | XP_006251016.1 | 4 | 1864 | |||
| XM_006250955.2 | XP_006251017.1 | 3 | 2052 | |||
| XM_008770152.1 | XP_008768374.1 | 3 | 1566 | |||
| GenBank mRNA | AY724525.1 | - | 3 | 801 | ||
| BC081995.1 | - | 2 | 778 | 7130530 | ||

Rbm47
Genomic Map