siRNA Details
| Gene Symbol: Sh3tc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107226.1 | NP_001100696.1 | 2, 3 | 308 | ||
| XM_006251125.2 | XP_006251187.1 | 3, 4 | 839 | |||
| XM_006251126.2 | XP_006251188.1 | 4, 5 | 650 | |||
| XM_006251127.2 | XP_006251189.1 | 3, 4 | 451 | |||
| XM_006251128.2 | XP_006251190.1 | 3, 4 | 1964 | |||
| XM_008770281.1 | XP_008768503.1 | 3, 4 | 1101 | |||
| XM_008770282.1 | XP_008768504.1 | 4, 5 | 525 | |||

Sh3tc1
Genomic Map