siRNA Details
| Gene Symbol: Nelfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008339.1 | NP_001008340.1 | 1 | 215 | ||
| GenBank mRNA | BC085948.1 | - | 1 | 215 | 7130899 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nelfa| Gene Symbol: Nelfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008339.1 | NP_001008340.1 | 1 | 215 | ||
| GenBank mRNA | BC085948.1 | - | 1 | 215 | 7130899 | |