siRNA Details
| Gene Symbol: Fam53a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107228.2 | NP_001100698.1 | 5 | 1195 | ||
| XM_006251262.2 | XP_006251324.1 | 5 | 1256 | |||
| XM_006251266.2 | XP_006251328.1 | 4 | 1157 | |||
| XM_006251267.2 | XP_006251329.1 | 4 | 1205 | |||
| XM_006251268.2 | XP_006251330.1 | 4 | 1130 | |||
| XM_006251270.2 | XP_006251332.1 | 4 | 670 | |||
| XM_008770286.1 | XP_008768508.1 | 4 | 1643 | |||
| XM_008770287.1 | XP_008768509.1 | 4 | 1647 | |||
| XM_008770288.1 | XP_008768510.1 | 4 | 674 | |||
| GenBank mRNA | BC168895.1 | - | 5 | 1195 | 7312953 | |

Fam53a
Genomic Map