siRNA Details
Gene Symbol: Fam53a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107228.2 | NP_001100698.1 | 6 | 1884 | ||
XM_006251262.2 | XP_006251324.1 | 6 | 1945 | |||
XM_006251266.2 | XP_006251328.1 | 5 | 1846 | |||
XM_006251267.2 | XP_006251329.1 | 5 | 1894 | |||
XM_006251268.2 | XP_006251330.1 | 5 | 1819 | |||
XM_006251270.2 | XP_006251332.1 | 5 | 1359 | |||
XM_008770286.1 | XP_008768508.1 | 5 | 2332 | |||
XM_008770287.1 | XP_008768509.1 | 5 | 2336 | |||
XM_008770288.1 | XP_008768510.1 | 5 | 1363 | |||
GenBank mRNA | BC168895.1 | - | 6 | 1884 | 7312953 |
