siRNA Details
| Gene Symbol: Sfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191682.1 | NP_001178611.1 | 15, 16 | 1621 | ||
| XM_006251285.2 | XP_006251347.1 | 16, 17 | 2020 | |||
| XM_006251287.2 | XP_006251349.1 | 17, 18 | 2076 | |||
| XM_006251288.2 | XP_006251350.1 | 16, 17 | 2020 | |||
| XM_006251289.2 | XP_006251351.1 | 16, 17 | 2020 | |||
| XM_008770294.1 | XP_008768516.1 | 15, 16 | 2117 | |||
| XM_008770295.1 | XP_008768517.1 | 10, 11 | 1903 | |||

Sfi1
Genomic Map