siRNA Details
Gene Symbol: Tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107238.1 | NP_001100708.1 | 3, 4 | 743 | ||
XM_006251444.2 | XP_006251506.1 | 4, 5 | 581 | |||
GenBank mRNA | BC168726.1 | - | 4, 5 | 553 | 6889842 | |
BC168735.1 | - | 4, 5 | 553 | 6891034 | ||
BC168766.1 | - | 4, 5 | 553 | 6890852 |
