siRNA Details
| Gene Symbol: Eif6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037352.1 | NP_001032429.1 | 5, 6 | 947 | ||
| XM_006235316.2 | XP_006235378.1 | 6, 7 | 840 | |||
| GenBank mRNA | BC105764.1 | - | 5, 6 | 947 | 7929717 | |
| FQ220694.1 | - | 5, 6 | 723 | |||
| FQ220773.1 | - | 6, 7 | 797 | |||

Eif6
Genomic Map