siRNA Details
| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107239.1 | NP_001100709.1 | 6 | 1144 | ||
| XM_008770320.1 | XP_008768542.1 | 8 | 1374 | |||
| XM_008770321.1 | XP_008768543.1 | 7 | 1302 | |||
| XM_008770322.1 | XP_008768544.1 | 6 | 723 | |||

Adcy1
Genomic Map