siRNA Details
Gene Symbol: Ehbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251565.1 | XP_006251627.1 | 8, 9 | 1291 | ||
XM_006251567.1 | XP_006251629.1 | 8, 9 | 1291 | |||
XM_008770445.1 | XP_008768667.1 | 8, 9 | 1067 | |||
XM_223664.7 | XP_223664.6 | 8, 9 | 1291 |
