siRNA Details
| Gene Symbol: Pus10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025278.1 | NP_001020449.1 | 2 | 478 | ||
| XM_008770428.1 | XP_008768650.1 | 2 | 422 | |||
| XM_008770429.1 | XP_008768651.1 | 2 | 422 | |||
| GenBank mRNA | BC093611.1 | - | 2 | 478 | 7307810 | |

Pus10
Genomic Map