siRNA Details
| Gene Symbol: Ddhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033066.1 | NP_001028238.1 | 6, 7 | 1515 | ||
| XM_006251769.2 | XP_006251831.1 | 8, 9 | 1706 | |||
| XM_006251770.2 | XP_006251832.1 | 7, 8 | 1685 | |||
| XM_006251771.2 | XP_006251833.1 | 8, 9 | 1706 | |||
| XM_006251772.2 | XP_006251834.1 | 7, 8 | 1604 | |||
| XM_006251773.2 | XP_006251835.1 | 6, 7 | 1583 | |||
| XM_006251774.2 | XP_006251836.1 | 7, 8 | 1603 | |||
| GenBank mRNA | BC103649.1 | - | 6, 7 | 1515 | 7314879 | |

Ddhd1
Genomic Map