siRNA Details
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013131.1 | NP_001013149.1 | 9, 10 | 1215 | ||
GenBank mRNA | BC091207.1 | - | 9, 10 | 1215 | 7381272 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013131.1 | NP_001013149.1 | 9, 10 | 1215 | ||
GenBank mRNA | BC091207.1 | - | 9, 10 | 1215 | 7381272 |