siRNA Details
| Gene Symbol: Zfhx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098803.1 | NP_001092273.1 | 5, 6 | 3220 | ||
| XM_008770677.1 | XP_008768899.1 | 4, 5 | 4995 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfhx2| Gene Symbol: Zfhx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098803.1 | NP_001092273.1 | 5, 6 | 3220 | ||
| XM_008770677.1 | XP_008768899.1 | 4, 5 | 4995 | |||