siRNA Details
| Gene Symbol: Thtpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007682.1 | NP_001007683.1 | 2 | 938 | ||
| XM_006251970.2 | XP_006252032.1 | 2 | 2909 | |||
| GenBank mRNA | AY065967.1 | - | 2 | 300 | ||
| BC081978.1 | - | 2 | 938 | 7128058 | ||

Thtpa
Genomic Map