siRNA Details
Gene Symbol: Slc25a37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013996.1 | NP_001014018.1 | 4 | 679 | ||
XM_006252266.1 | XP_006252328.1 | 5 | 851 | |||
GenBank mRNA | BC082071.1 | - | 4 | 679 | 7132913 | |
FQ227341.1 | - | 4 | 620 | |||
FQ227813.1 | - | 1 | 14 | |||
FQ230701.1 | - | 3 | 336 | |||
FQ231671.1 | - | 3 | 150 | |||
FQ231945.1 | - | 3 | 167 | |||
FQ233331.1 | - | 4 | 579 |
