siRNA Details
| Gene Symbol: Kbtbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012045.2 | NP_001012045.2 | 1 | 838 | ||
| GenBank mRNA | BC086326.1 | - | 677 | 7114652 | ||
| Gene Symbol: Kbtbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252370.2 | XP_006252432.1 | 1100 | |||

Kbtbd6
Genomic Map