siRNA Details
| Gene Symbol: Klf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107281.1 | NP_001100751.1 | 5 | 956 | ||
| XM_006252408.2 | XP_006252470.1 | 5 | 907 | |||
| XM_006252409.2 | XP_006252471.1 | 5 | 902 | |||
| XM_006252410.2 | XP_006252472.1 | 5 | 973 | |||
| XM_006252413.2 | XP_006252475.1 | 4 | 755 | |||
| XM_006252415.2 | XP_006252477.1 | 4 | 752 | |||
| XM_008770870.1 | XP_008769092.1 | 4 | 2709 | |||
| XM_008770871.1 | XP_008769093.1 | 6 | 919 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010636, Y14295, AB221575, BC067408.

Klf12
Genomic Map