siRNA Details
| Gene Symbol: Spry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012046.1 | NP_001012046.1 | 2 | 676 | ||
| XM_006252438.2 | XP_006252500.1 | 2 | 970 | |||
| GenBank mRNA | BC089116.1 | - | 2 | 676 | 7320532 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spry2| Gene Symbol: Spry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012046.1 | NP_001012046.1 | 2 | 676 | ||
| XM_006252438.2 | XP_006252500.1 | 2 | 970 | |||
| GenBank mRNA | BC089116.1 | - | 2 | 676 | 7320532 | |