siRNA Details
| Gene Symbol: Slitrk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107284.1 | NP_001100754.1 | 1 | 864 | ||
| XM_006252444.2 | XP_006252506.1 | 2 | 1469 | |||
| XM_006252445.2 | XP_006252507.1 | 2 | 1132 | |||
| XM_006252446.2 | XP_006252508.1 | 2 | 942 | |||
| XM_006252447.2 | XP_006252509.1 | 2 | 1238 | |||

Slitrk5
Genomic Map