siRNA Details
| Gene Symbol: Tgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107286.1 | NP_001100756.1 | 4, 5 | 397 | ||
| GenBank mRNA | BC166991.1 | - | 1, 2 | 67 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgds| Gene Symbol: Tgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107286.1 | NP_001100756.1 | 4, 5 | 397 | ||
| GenBank mRNA | BC166991.1 | - | 1, 2 | 67 | ||