siRNA Details
| Gene Symbol: Clybl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100685.1 | NP_001094155.1 | 7, 8 | 935 | ||
| XM_006252508.2 | XP_006252570.1 | 6, 7 | 764 | |||
| GenBank mRNA | BC088243.1 | - | 7, 8 | 915 | ||
| FQ214276.1 | - | 6, 7 | 720 | |||

Clybl
Genomic Map