siRNA Details
| Gene Symbol: Duxbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252678.2 | XP_006252740.1 | 6 | 3583 | ||
| XM_006252679.2 | XP_006252741.1 | 7 | 1584 | |||
| XM_008771030.1 | XP_008769252.1 | 4 | 1075 | |||
| XM_008771031.1 | XP_008769253.1 | 5 | 4258 | |||
| XM_224643.6 | XP_224643.1 | 6 | 2259 | |||

Duxbl1
Genomic Map