siRNA Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107295.1 | NP_001100765.1 | 6 | 817 | ||
| XM_006252623.2 | XP_006252685.1 | 6 | 918 | |||
| XM_006252624.2 | XP_006252686.1 | 7 | 1025 | |||
| XM_006252625.2 | XP_006252687.1 | 7 | 1011 | |||
| XM_006252626.2 | XP_006252688.1 | 6 | 904 | |||
| XM_008771000.1 | XP_008769222.1 | 7 | 1106 | |||

Oxnad1
Genomic Map